missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSPP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11931-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a centrosome and spindle pole associated protein. The encoded protein plays a role in cell-cycle progression and spindle organization, regulates cytokinesis, interacts with Nephrocystin 8 and is required for cilia formation. Mutations in this gene result in primary cilia abnormalities and classical Joubert syndrome. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Specifications
| CSPP1 | |
| Polyclonal | |
| Unconjugated | |
| CSPP1 | |
| CSPP, CSPP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 211660, 362472, 79848 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B2RX88, Q1MSJ5 | |
| Cspp1 | |
| CSPP1 Fusion Protein Ag2521 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction