Learn More
CTCF Antibody (CL0307) [Alexa Fluor™ 700], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52911AF700
Product Code. 30292815
  • 770.00€ / 0.10mL
Description
CTCF Monoclonal antibody specifically detects CTCF in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CTCF
Monoclonal
Alexa Fluor 700
11 zinc finger transcriptional repressor, 11-zinc finger protein, CCCTC-binding factor, CCCTC-binding factor (zinc finger protein), CTCFL paralog, transcriptional repressor CTCF
This antibody was developed using a recombinant protein derived from P49711, with the exact immunogen sequence remaining proprietary.
0.1 mL
Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
10664
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL0307