missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRB1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24939-1-AP-20UL
This item is not returnable.
View return policy
Description
Preferential cleavage: Tyr--Xaa, Trp--Xaa, Phe--Xaa, Leu--Xaa.Specifications
| CTRB1 | |
| Polyclonal | |
| Unconjugated | |
| CTRB1 | |
| chymotrypsinogen B1, CTRB, CTRB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1504, 24291 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P07338, P17538 | |
| Ctrb1 | |
| CTRB1 Fusion Protein Ag8604 | |
| 20 μL | |
| Primary | |
| Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction