Learn More
Sartorius™ Cubis™ MSE Analytical Balance
Designed for weighing with maximum accuracy, without complex workflows.
Brand:  Sartorius™ MSE225P-1CE-DU
Additional Details : Weight : 6.80000kg
 View more versions of this product
Product Code. 15460563
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Description
Description
  • Large, high-contrast LCD
  • Simple menu guidance with short-text prompts
  • Clear structured keys
Specifications
Specifications
Black and White LCD
0.01, 0.02, 0.05 mg
Automatic
30 mg