missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUEDC2 Antibody (003), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89902CL1
This item is not returnable.
View return policy
Description
CUEDC2 Monoclonal antibody specifically detects CUEDC2 in Human samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| CUEDC2 | |
| Monoclonal | |
| CoraFluor 1 | |
| bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CUEDC2. | |
| 0.1 mL | |
| Signal Transduction | |
| 79004 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 003 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction