missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUGBP2 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-204PE
This item is not returnable.
View return policy
Description
CUGBP2 Polyclonal antibody specifically detects CUGBP2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| CUGBP2 | |
| Polyclonal | |
| Western Blot | |
| BRUNOL3ELAV-type RNA-binding protein 3, Bruno-like protein 3, CELF-2, CUG triplet repeat RNA-binding protein 2, CUG triplet repeat, RNA binding protein 2, CUG triplet repeat, RNA-binding protein 2, CUG-BP- and ETR-3-like factor 2, CUGBP Elav-like family member 2, CUGBP, Elav-like family member 2, CUG-BP2, CUGBP2hNAPOR, Etr-3, ETR3RNA-binding protein BRUNOL-3, NAPOR, NAPOR-2, Neuroblastoma apoptosis-related RNA-binding protein | |
| C-terminus of human and mouse ETR-3 | |
| 0.1 mL | |
| Lipid and Metabolism, Transcription Factors and Regulators | |
| 10659 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction