missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL4A Mouse anti-Human, Mouse, Non-human primate, Porcine, Rat, Clone: 1A7F12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66038-1-IG-20UL
This item is not returnable.
View return policy
Description
CUL4A (Cullin 4A) is the ubiquitin ligase component of a multimeric complex involved in the degradation of DNA damage-response proteins.Specifications
| CUL4A | |
| Monoclonal | |
| 0.76 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13619, Q3TCH7 | |
| Cul4a | |
| CUL4A Fusion Protein Ag18089 | |
| 20 μL | |
| Primary | |
| Non-human Primate, Rat, Human, Mouse, Pig | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence | |
| 1A7F12 | |
| Unconjugated | |
| CUL4A | |
| CUL 4A, CUL4A, cullin 4A | |
| Mouse | |
| Protein G | |
| RUO | |
| 100517912, 361181, 8451, 99375 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction