Learn More
Ambion™ Custom mirVana™ miRNA Mimic
Product Code. 15551124
Change view
Click to view available options
Quantity:
20 nmol
250 nmol
5 nmol
Unit Size:
20nmol
250nmol
5nmol
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15551124 250 nmol 250nmol
15531124 5 nmol 5nmol
15541124 20 nmol 20nmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15551124 Supplier Ambion™ Supplier No. 4464071
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Custom mirVana™ miRNA Mimic
miRNA mimics are small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity. Use our GeneAssist™ miRNA Workflow Builder to design both mimics and inhibitors for any miRNA sequence.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Customer-Defined mirVana™ miRNA mimic, 250nmol, Lowest Off-Target Effects and Highest Potency, miRNA mimics are small, Chemically Modified Double-Stranded RNAs that mimic Endogenous miRNAs and Enable miRNA Functional Analysis by Up-Regulation of miRNA Activity
Product Type Customer-defined mirVana™ miRNA mimic
Quantity 250 nmol