Learn More
Ambion™ Custom RNA, Stealth, Desalted
Product Code. 10459213
Change view
Click to view available options
Quantity:
100 nmol
2 nmol
25 nmol
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10459213 25 nmol Each
10276583 2 nmol Each
10184923 100 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10459213 Supplier Ambion™ Supplier No. 100001854
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Custom RNA, Stealth, Desalted
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description RNA DSL In-vivo, 25nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation
Product Type RNA DSL In-vivo
Quantity 25 nmol