missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL13/BLC/BCA-1 Antibody (CL10543) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07988B
This item is not returnable.
View return policy
Description
CXCL13/BLC/BCA-1 Monoclonal antibody specifically detects CXCL13/BLC/BCA-1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| CXCL13/BLC/BCA-1 | |
| Monoclonal | |
| Biotin | |
| ANGIE, ANGIE2, B cell-attracting chemokine 1, B lymphocyte chemoattractant, BCA1B-cell chemoattractant, BCA-1CXC chemokine BLC, B-cell-homing chemokine (ligand for Burkitt's lymphoma receptor-1), BLCSmall-inducible cytokine B13, BLR1L, B-lymphocyte chemoattractant, chemokine (C-X-C motif) ligand 13, chemokine (C-X-C motif) ligand 13 (B-cell chemoattractant), C-X-C motif chemokine 13, SCYB13B-cell-attracting chemokine 1, small inducible cytokine B subfamily (Cys-X-Cys motif), member 13 (B-cellchemoattractant) | |
| This antibody was developed using a synthetic peptide derived from O43927, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research, Signal Transduction | |
| 10563 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10543 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction