Learn More
PeproTech CXCL6 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
219.00€ - 3435.00€
Specifications
| Antigen | CXCL6 |
|---|---|
| Concentration | 0.1-1.0 mg/mL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17844263
![]() |
PeproTech
500-P120BT-25UG |
25 μg |
219.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17814743
![]() |
PeproTech
500-P120BT-50UG |
50 μg |
275.00€
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17883893
|
Invitrogen
500-P120BT-1MG |
2 x 500 μg | |||||||
|
30054779
|
PeproTech
500-P120BT-500UG |
500 μg |
1836.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AA Sequence of recombinant protein: VLTELRCTCL RVTLRVNPKT IGKLQVFPAG PQCSKVEVVA SLKNGKQVCL DPEAPFLKKV IQKILDSGNK KN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GCP-2 (CXCL6). Anti-Human GCP-2 (CXCL6)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human GCP-2 (CXCL6) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human GCP-2 (CXCL6) (500-P120) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human GCP-2 (CXCL6). Western Blot: To detect hGCP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hGCP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P120BT-1MG will be provided as 2 x 500 μg
CXCL6 is a chemotactic for neutrophil granulocytes. It signals through binding and activation of its receptors (CXCR1 and CXCR2). In addition to its chemotactic and angiogenic properties, it has strong antibacterial activity against Gram-positive and Gram-negative bacteria (90-fold-higher when compared to CXCL5 and CXCL7).Specifications
| CXCL6 | |
| ELISA, Western Blot | |
| Biotin | |
| Rabbit | |
| Human | |
| P80162 | |
| 6372 | |
| E.coli-derived Recombinant Human GCP-2 (CXCL6). | |
| Antigen affinity chromatography | |
| CXCL6 |
| 0.1-1.0 mg/mL | |
| Polyclonal | |
| Lyophilized | |
| RUO | |
| PBS with no preservative | |
| C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); chemokine alpha 3; CKA3; CKA-3; CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine 6; C-X-C motif chemokine ligand 6; CXCL; Cxcl5; CXCL6; Cytokine LIX; GCP2; GCP-2; granulocyte chemotactic protein 2; Scyb5; SCYB6; small inducible cytokine subfamily B (Cys-X-Cys), member 6 (granulocyte chemotactic protein 2); Small inducible cytokine subfamily B (Cys-X-Cys), member b; small-inducible cytokine B5; Small-inducible cytokine B6; Small-inducible cytokine B6, N-processed variant 1; Small-inducible cytokine B6, N-processed variant 2; Small-inducible cytokine B6, N-processed variant 3 | |
| CXCL6 | |
| Primary | |
| -20°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.