Learn More
Invitrogen™ CXCL9 (MIG) Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558542
 View more versions of this product
Product Code. 30282990
  • 1204.00€ / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term Novimmune patent anti-CXCL9.
MIG, also known as CXCL9, is a T-cell chemoattractant inducible by gamma interferon that is a member of the CXC chemokine family of cytokines.This gene is mapped to 4q21. It is noted that, although the best-described activities of the chemokines are as chemotactic factors, chemokines also have an effect on T-cell activation, angiogenesis, and HIV infection. While most CXC chemokines are chemotactic for neutrophils, MIG and INP10 are unusual and similar in being CXC chemokines that are chemotactic for lymphocytes and inactive in neutrophils.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL9 (MIG) Biosimilar
Recombinant Monoclonal
Unconjugated
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A