missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR2/IL-8RB Antibody (5E8-C7-F10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
285.00€ - 628.00€
Specifications
| Antigen | CXCR2/IL-8 RB |
|---|---|
| Clone | 5E8-C7-F10 |
| Concentration | 0.5 mg/mL |
| Dilution | Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243961
|
Novus Biologicals
NBP1-43338-0.025MG |
0.025 mg |
285.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18083514
|
Novus Biologicals
NBP1-43338-0.1MG |
0.1 mg |
487.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18268692
|
Novus Biologicals
NBP1-43338-0.5MG |
0.5 mg |
628.00€
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CXCR2/IL-8 RB | |
| 0.5 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| beta, CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B | |
| CXCR2 | |
| IgG1 κ | |
| Affinity Purified |
| 5E8-C7-F10 | |
| Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100 | |
| Monoclonal | |
| Mouse | |
| Angiogenesis, Cancer, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| 3579 | |
| The immunogen for this antibody was CD182. | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title