missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cy5-Fuc Labeled M1N1f
Description
- Excitation at 649 nm and emission at 671 nm
- The fluorescent dye Cy5 is conjugated to the C6 position of the core-6 fucose
- Can be separated on 15 to 17% SDS-PAGE and directly visualized as a single band under a fluorescent imager
- Linear response range for Cy5-labeled glycans can be from 10 fmol to 100 pmol, depending on the sensitivity of detection
- Store at <-20°C. Good for 6 months from date of receipt.
Specifications
Specifications
| Quantity | 50 pmol |
| Target | M1N1f |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?