missing translation for 'onlineSavingsMsg'
Learn More

Cyanamide, TRC

Product Code. 30361589
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30361589 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361589 Supplier Toronto Research Chemicals Supplier No. TRCC95250050G

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyanamide
CAS 420-04-2
Purity Grade Notes GC
Molecular Formula C H2 N2
Quantity 50 g
Synonym Alzogur, Amidocyanogen, Carbamonitrile, Carbimide, Cyanamide F 1000, Cyanoamine, Cyanogen nitride, Cyanogenamide, Deurbraak, Dormex, Dyhard VP 111, F 1000, Hydrogen cyanamide, N-Cyanoamine, NSC 24133, TsAKS
InChI Formula InChI=1S/CH2N2/c2-1-3/h2H2
SMILES NC#N
IUPAC Name cyanamide
Molecular Weight (g/mol) 42.04
Formula Weight 42.0218
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.