missing translation for 'onlineSavingsMsg'
Learn More

Cyanide Standard Solution, Ion HIQU, Chem-Lab

Product Code. 11969042
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11969042 500 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11969042 Supplier Chem-Lab Supplier No. CL01.0371.0100
  • ONLINE EXCLUSIVE PRICE
    73.00€
    35.40€ / 100mL
    Save 37.60€
    51% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Used to dissolve metals and their ores. Cyanide Standard Solution, Ion HIQU, Chem-Lab is comprised of 2.503g potassium cyanide per liter of water. The solution contains 1000μg/mL of CN- (±2μg, at -20°C).

Cyanide standard solution is used to dissolve metals and their ores. It is also used in clinical chemistry and in waste water treatment facilities to determine the concentration of dissolved and particulate materials. Potassium cyanide is highly toxic. In aqueous solution, KCN is dissociated into hydrated potassium (K+) ions and cyanide (CN−) ions.

Specifications

CAS 151-50-8
Density 1.00g/mL
Molecular Formula CKN
Concentration 1000 μg/mL of CN− (±2μg, at −20°C)
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
SMILES [C-]#N.[K+]
IUPAC Name potassium;cyanide
Molecular Weight (g/mol) 65.116
PubChem CID 9032
Quantity 500 mL
Formula Weight 65.12g/mol
Concentration or Composition (by Analyte or Components) 2.503g KCN per 1L H2O
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyanide standard solution 1000 µg/ml for IC (Ion HIQU)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contain: potassium cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.