missing translation for 'onlineSavingsMsg'
Learn More

Cyanoacetamide, TRC

Product Code. 17970915
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17970915 250 g 250g
17933515 25 g 25g
17965005 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17970915 Supplier Toronto Research Chemicals Supplier No. TRCC987460250G
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-91-5
Molecular Formula C3 H4 N2 O
Molecular Weight (g/mol) 84.0767
Synonym 2-Cyanoacetamide, Cyanacetamide, Cyanoacetic Acid Amide, Cyanoiminoacetic Acid, Malonamide Nitrile, Malonamonitrile, NSC 6285, NSC 8948, Nitrilomalonamide, α-Cyanoacetamide
IUPAC Name 2-cyanoacetamide
SMILES NC(=O)CC#N

Specifications

Quantity 250 g
InChI Formula InChI=1S/C3H4N2O/c4-2-1-3(5)6/h1H2,(H2,5,6)
Formula Weight 84.0324
Chemical Name or Material Cyanoacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.