missing translation for 'onlineSavingsMsg'
Learn More

Cyantraniliprole, TRC

Product Code. 30004915
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004915 250 mg 250mg
17902155 100 mg 100mg
17984925 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004915 Supplier Toronto Research Chemicals Supplier No. TRCC987580250MG
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 736994-63-1
Molecular Formula C19 H14 Br Cl N6 O2
Molecular Weight (g/mol) 473.71
Synonym 1H-Pyrazole-5-carboxamide, 3-bromo-1-(3-chloro-2-pyridinyl)-N-[4-cyano-2-methyl-6-[(methylamino)carbonyl]phenyl]-, 3-Bromo-1-(3-chloro-2-pyridinyl)-N-[4-cyano-2-methyl-6-[(methylamino)carbonyl]phenyl]-1H-pyrazole-5-carboxamide, 3-Bromo-1-(3-chloropyridin-2-yl)-N-[4-cyano-2-methyl-6-[N-methylcarbamoyl]phenyl]-1H-pyrazole-5-carboxamide, Benevia, Cyantraniliprole, Cyazypyr, DPX-HGW 86, Exirel, Exirel (insecticide), HGW 20SC, HGW 86, SYN 545377, Verimark
IUPAC Name 5-bromo-2-(3-chloropyridin-2-yl)-N-[4-cyano-2-methyl-6-(methylcarbamoyl)phenyl]pyrazole-3-carboxamide
SMILES CNC(=O)c1cc(cc(C)c1NC(=O)c2cc(Br)nn2c3ncccc3Cl)C#N

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H14BrClN6O2/c1-10-6-11(9-22)7-12(18(28)23-2)16(10)25-19(29)14-8-15(20)26-27(14)17-13(21)4-3-5-24-17/h3-8H,1-2H3,(H,23,28)(H,25,29)
Formula Weight 472.005
Chemical Name or Material Cyantraniliprole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.