missing translation for 'onlineSavingsMsg'
Learn More

Cyclamen Alcohol, TRC

Product Code. 30179678
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179678 1 g 1g
30179064 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179678 Supplier Toronto Research Chemicals Supplier No. TRCC9876551G
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4756-19-8
Molecular Formula C13 H20 O
Molecular Weight (g/mol) 192.2973
Synonym 1-Propanol, 3-p-cumenyl-2-methyl- (6CI,7CI,8CI), β-Methyl-4-(1-methylethyl)benzenepropanol, 2-Methyl-3-(4-propan-2-ylphenyl)propan-1-ol, 2-Methyl-3-[4-(propan-2-yl)phenyl]propan-1-ol, 3-(4-Isopropylphenyl)-2-methyl-1-propanol, 3-p-Cumenyl-2-methyl-1-propanol, Cyclamen alcohol, p-Isopropyl-β-methylhydrocinnamic alcohol, Cyclamen Alcohol
IUPAC Name 2-methyl-3-(4-propan-2-ylphenyl)propan-1-ol
SMILES CC(CO)Cc1ccc(cc1)C(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H20O/c1-10(2)13-6-4-12(5-7-13)8-11(3)9-14/h4-7,10-11,14H,8-9H2,1-3H3
Formula Weight 192.1514
Chemical Name or Material Cyclamen Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.