Learn More
Cyclaniliprole, TRC
Product Code. 30363978
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30363978 5 mg 5mg
30353433 50 mg 50mg
30363125 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30363978 Supplier Toronto Research Chemicals Supplier No. TRCC9824905MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1031756-98-5
Molecular Formula C21 H17 Br2 Cl2 N5 O2
Molecular Weight (g/mol) 602.11
Synonym 3-Bromo-N-(2-bromo-4-chloro-6-(1-cyclopropylethylcarbamoyl)phenyl)-1-(3-chloropyridin-2-yl)-1H-pyrazole-5-carboxamide, 3-Bromo-N-[2-bromo-4-chloro-6-[[(1-cyclopropylethyl)amino]carbonyl]phenyl]-1-(3-chloropyridin-2-yl)-1H-pyrazole-5-carboxamide, Cyclaniliprole, IKI 3106, N-[2-Bromo-4-chloro-6-[[α-methyl(cyclopropylmethyl)amino]carbonyl]phenyl]-3-bromo-1-(3-chloro-2-pyridinyl)-1H-pyrazole-5-carboxamide, 1H-Pyrazole-5-carboxamide, 3-bromo-N-[2-bromo-4-chloro-6-[[(1-cyclopropylethyl)amino]carbonyl]phenyl]-1-(3-chloro-2-pyridinyl)-, Cyclaniliprole
IUPAC Name 5-bromo-N-[2-bromo-4-chloro-6-(1-cyclopropylethylcarbamoyl)phenyl]-2-(3-chloropyridin-2-yl)pyrazole-3-carboxamide
SMILES CC(NC(=O)c1cc(Cl)cc(Br)c1NC(=O)c2cc(Br)nn2c3ncccc3Cl)C4CC4
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C21H17Br2Cl2N5O2/c1-10(11-4-5-11)27-20(31)13-7-12(24)8-14(22)18(13)28-21(32)16-9-17(23)29-30(16)19-15(25)3-2-6-26-19/h2-3,6-11H,4-5H2,1H3,(H,27,31)(H,28,32)
Formula Weight 598.9126
Chemical Name or Material Cyclaniliprole