missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclic AMP Antibody (9H4C4), Novus Biologicals™
Mouse Monoclonal Antibody
187.00€
Specifications
| Antigen | Cyclic AMP |
|---|---|
| Clone | 9H4C4 |
| Dilution | ELISA 1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Cyclic AMP Monoclonal specifically detects Cyclic AMP in Non-species specific samples. It is validated for ELISA.Specifications
| Cyclic AMP | |
| ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| 3' 5' cyclic adenosine monophosphate, Cyclic adenosine monophosphate, Cyclic AMP | |
| IgG1 | |
| Unpurified |
| 9H4C4 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Non-species specific | |
| Synthetic peptide of cAMP, conjugated to KLH. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title