missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Cyclic Pifithrin-Alpha (Hydrobromide)

Product Code. p-7217825
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17100022 5 mg 5mg
17110022 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17100022 Supplier STEMCELL Technologies Supplier No. 72062

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p53 Inhibitor

  • Cyclic Pifthrin-Alpha is a cell-permeable and reversible inhibitor of p53-mediated apoptosis and p53-dependent gene transcription
  • It is a more stable and less cytotoxic analog of the non-cyclic form of pifithrin-alpha
  • Cyclic Pifithrin-Alpha has also been reported to activate the aryl hydrocarbon receptor
  • (Fernandez-Cruz et al.), Gary & Jensen, Komarov et al.)
  • Increases efficiency of reprogramming mouse embryonic fibroblasts to induced pluripotent stem cells (Liao et al.)
  • Reduces UV-induced apoptosis of mouse embryonic stem cells (Qin et al.)
  • Increases the numbers of mouse hematopoietic stem and progenitor cells in vivo and in vitro, also decreases the radiation-induced death of these cells (Leonova et al.)

Specifications

CAS 511296-88-1
Synonym Cyclic PFT-α; Cyclic Pifithrin-α; PFT-β; Pifithrin-β
Quantity 5 mg
Molecular Formula C16H16N2S · HBr
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.