missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cyclin A2 Recombinant Rabbit Monoclonal Antibody (23GB1875), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
217.00€ - 590.00€
Specifications
| Antigen | Cyclin A2 |
|---|---|
| Clone | 23GB1875 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089803
![]() |
Invitrogen™
MA554060 |
100 μL |
590.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089496
|
Invitrogen™
MA554061 |
20 μL |
217.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclin A2 Recombinant Monoclonal antibody specifically detects Cyclin A2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Cyclin A2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P20248, P51943 | |
| 114494, 12428, 890 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1875 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AA408589; Ccn1; Ccn-1; CCNA; CCNA2; ccna2.S; Cyca; CycA2; Cyclin A; cyclin A2; Cyclin A2 (Cyclin A); cyclin A2 S homeolog; Cyclin A-3; cyclin-A; Cyclin-A2; XELAEV_18009326mg | |
| A synthesized peptide derived from human Cyclin A2 (1-250AA). | |
| Primary | |
| -20°C | |
| CCNA2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title