missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D1 Antibody (DCS-6) - IHC-Prediluted, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44584
This item is not returnable.
View return policy
Description
Cyclin D1 Monoclonal antibody specifically detects Cyclin D1 in Human, Mouse, Rat, Monkey samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Cyclin D1 | |
| Monoclonal | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DCS-6 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 0.5 - 1.0 μg/mL | |
| B-cell lymphoma 1 protein, BCL-1, BCL-1 oncogene, BCL1D11S287E, cyclin D1, cyclin D1 (PRAD1: parathyroid adenomatosis 1), G1/S-specific cyclin D1, G1/S-specific cyclin-D1, PRAD1 oncogene, PRAD1B-cell CLL/lymphoma 1, U21B31 | |
| Human full length recombinant cyclin D1 protein (Uniprot: P24385) | |
| 7 mL | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, mTOR Pathway, Stem Cell Markers, Wnt Signaling Pathway | |
| 595 | |
| Store at 4°C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction