Learn More
Cyclobutyl methyl ketone, 97%
Product Code. 11352196
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11352196 1 g 1g
11362196 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11352196 Supplier Thermo Scientific Alfa Aesar Supplier No. L03521.03
Request Bulk or Custom Format
CAS: 3019-25-8 | C6H10O | 98.145 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3019-25-8
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00037148
InChI Key JPJOOTWNILDNAW-UHFFFAOYSA-N
Synonym cyclobutyl methyl ketone, 1-cyclobutylethan-1-one, acetylcyclobutane, ethanone, 1-cyclobutyl, ketone, cyclobutyl methyl, methyl cyclobutyl ketone, cyclobutylmethylketone, acetylcyclobutan, cyclobutylethanone, cyclobutane, acetyl
PubChem CID 76398
IUPAC Name 1-cyclobutylethanone
SMILES CC(=O)C1CCC1
Specifications
Density 0.9
Boiling Point 134°C to 135°C
Flash Point 28°C (82°F)
Refractive Index 1.432
Quantity 1 g
UN Number UN1224
Beilstein 1924022
Formula Weight 98.15
Percent Purity 97%
Chemical Name or Material Cyclobutyl methyl ketone