missing translation for 'onlineSavingsMsg'
Learn More

γ-Cyclodextrin, MP Biomedicals™

Product Code. 11434512 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434512 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434512 Supplier MP Biomedicals Supplier No. 02190054.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

γ-Cyclodextrin, MP Biomedicals is reported to be useful as a fluorescent enhancer for chemical and clinical assays.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17465-86-0
Molecular Formula C48H80O40
Molecular Weight (g/mol) 1297.13
MDL Number MFCD00009595,MFCD00149574
InChI Key GDSRMADSINPKSL-UHFFFAOYNA-N
Synonym cyclooctaamylose, schardinger gamma-dextrin
PubChem CID 101136809
IUPAC Name 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Melting Point 257°C to 265°C (decomposition)(literature)
Quantity 500 mg
Solubility Information Soluble in water, 1M NH4OH (50mg/mL)
Formula Weight 1297.1
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.