missing translation for 'onlineSavingsMsg'
Learn More

Cycloheptene, 96%, stab. with 0.1% BHT

Product Code. 11462220
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462220 5 g 5g
11472220 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462220 Supplier Thermo Scientific Alfa Aesar Supplier No. H29062.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-92-2 | C7H12 | 96.173 g/mol

Cycloheptene was used in one-pot synthesis of 1,2/3-triols from the allylichydroperoxides catalyzed by zeolite-confined osmium(0) nanoclusters. It is a raw material in organic chemistry and a monomer in polymer synthesis. Copper (1) triflate catalyzes the photodimerization of the simple nonconjugated unstrained olefins, cyclopentene, cyclohexene, and cycloheptene, but not cyclooctene or acyclic olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cycloheptene was used in one-pot synthesis of 1,2/3-triols from the allylichydroperoxides catalyzed by zeolite-confined osmium(0) nanoclusters. It is a raw material in organic chemistry and a monomer in polymer synthesis. Copper (1) triflate catalyzes the photodimerization of the simple nonconjugated unstrained olefins, cyclopentene, cyclohexene, and cycloheptene, but not cyclooctene or acyclic olefins.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids. May react exothermically with reducing agents to release hydrogen gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-92-2
Molecular Formula C7H12
Molecular Weight (g/mol) 96.173
MDL Number MFCD00004156
InChI Key ZXIJMRYMVAMXQP-UHFFFAOYSA-N
Synonym cis-cycloheptene, z-cycloheptene, cyclohept-1-ene, 1-cycloheptene, z, trans-cyclohepten, e-cycloheptene, 1-cycloheptene #, cycloheptene, cycloheptene, pure
PubChem CID 12363
IUPAC Name cycloheptene
SMILES C1CCC=CCC1

Specifications

Density 0.833
Boiling Point 112°C to 115°C
Flash Point −7°C (19°F)
Refractive Index 1.458
Quantity 5 g
UN Number UN2242
Solubility Information Insoluble in water.
Formula Weight 96.17
Percent Purity 96%
Chemical Name or Material Cycloheptene, stabilized with 0.1% BHT

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.