missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarboxaldehyde, 97%

Product Code. 15433257
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15433257 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15433257 Supplier Thermo Scientific Alfa Aesar Supplier No. B23837.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2043-61-0 | C7H12O | 112.17 g/mol

Cyclohexanecarboxaldehyde is used for the synthesis of poly(vinyl cyclohexanal). It was used as model substrate in the synthesis of propargylamines using Fe3O4 nanoparticles as catalyst. It was employed in an aldol reaction with ethyl diazoacetate catalyzed by basic, nanoparticulate magnesium oxide leading to α-diazo-β-hydroxyesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarboxaldehyde is used for the synthesis of poly(vinyl cyclohexanal). It was used as model substrate in the synthesis of propargylamines using Fe3O4 nanoparticles as catalyst. It was employed in an aldol reaction with eth
TRUSTED_SUSTAINABILITY

Specifications

CAS 2043-61-0
Melting Point 34°C to 35°C
Density 0.93
Boiling Point 161°C to 163°C
Flash Point 40°C (105°F)
Molecular Formula C7H12O
Refractive Index 1.45
MDL Number MFCD00001457
Quantity 25 g
UN Number UN1989
Beilstein 878306
Sensitivity Air and light sensitive
Synonym cyclohexanecarboxaldehyde, cyclohexanal, formylcyclohexane, cyclohexanealdehyde, cyclohexanaldehyde, cyclohexylcarboxaldehyde, 1-formylcyclohexane, hexahydrobenzaldehyde, cyclohexylformaldehyde, cyclohexane formaldehyde
Solubility Information Soluble in chloroform,ether,alcohol. Slightly soluble in water.
InChI Key KVFDZFBHBWTVID-UHFFFAOYSA-N
SMILES O=CC1CCCCC1
IUPAC Name cyclohexanecarbaldehyde
Molecular Weight (g/mol) 112.17
PubChem CID 16275
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material Cyclohexanecarboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.