Learn More
Cyclohexanecarboxylic acid, 98%
Product Code. 10224860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10224860 1 kg Glass bottle 1kg
10173380 25 g Glass bottle 25g
10193380 100 g Glass bottle 100g
10214910 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10224860 Supplier Thermo Scientific Acros Supplier No. 146560010
Request Bulk or Custom Format
CAS: 98-89-5 | C7H12O2 | 128.17 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98-89-5
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001461
InChI Key NZNMSOFKMUBTKW-UHFFFAOYSA-N
Synonym hexahydrobenzoic acid, carboxycyclohexane, cyclohexanoic acid, cyclohexylcarboxylic acid, cyclohexylmethanoic acid, cyclohexylformic acid, benzoic acid, hexahydro, cyclohexancarbonsaeure, cyclohexane-1-carboxylate, cyclohexanecarboxylicacid
PubChem CID 7413
ChEBI CHEBI:36096
IUPAC Name cyclohexanecarboxylic acid
SMILES C1CCC(CC1)C(=O)O
Specifications
Melting Point 30.0°C to 32.0°C
Color White
Density 1.0300g/mL
Boiling Point 232.0°C to 233.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H11CO2H
Refractive Index 1.4600 to 1.4620
Quantity 1 kg
Beilstein 09, 7
Merck Index 15, 2717
Specific Gravity 1.03
Solubility Information Solubility in water: 0.201g/100mL (15 c). Other solubilities: soluble in alcohol and ether
Formula Weight 128.17
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material Cyclohexanecarboxylic acid, 98+%