Learn More
Retsch™ Cyclone Unit
Retsch™ Cyclone Unit is an optional dust extraction accessory for use with a rotor beater or cross beater mill.
Brand:  Retsch™ 22.935.0020
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15924510
  • 4850.00€ / Each
Estimated Shipment: 08-08-2023
to see stock.
Add to basket
Description
Description
  • Includes a 5 L collecting receptacle
  • To be used with industrial vacuum cleaner
Specifications
Specifications
Cyclone Suction Combination Unit
Rotor Beater Mill SR 300
5 L collection receptacle