missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctane, 99+%

Product Code. 11338106
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338106 250 mL 250mL
11328106 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338106 Supplier Thermo Scientific Alfa Aesar Supplier No. L15000.AK
  • ONLINE EXCLUSIVE PRICE
    175.44€
    144.65€ / 250mL
    Save 30.79€
    17% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 292-64-8 | C8H16 | 112.216 g/mol

Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctane is used as solvent, reagents and synthetic organic intermediates and it can be used to produce cyclooctanone. It is also used as an intermediate in production of plastics, fibers, adhesives and coatings.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 292-64-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00004162
InChI Key WJTCGQSWYFHTAC-UHFFFAOYSA-N
Synonym octamethylene, cyclooctan, unii-kkz3kbs654, kkz3kbs654, cyclooctane, acmc-209h7x, cambridge id 6337558, cambridge id 6511221, cambridge id 6544558, cyclooctane, analytical standard
PubChem CID 9266
IUPAC Name cyclooctane
SMILES C1CCCCCCC1

Specifications

Melting Point 12°C to 14°C
Density 0.835
Boiling Point 150°C to 152°C
Flash Point 28°C (82°F)
Refractive Index 1.458
Quantity 250 mL
UN Number UN3295
Beilstein 1900349
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 112.22
Percent Purity ≥99%
Chemical Name or Material Cyclooctane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.