missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclooctane-d16, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Cyclooctane-d16 |
| CAS | 92204-03-0 |
| Molecular Formula | C8 D16 |
| Quantity | 0.5 g |
| Synonym | Cyclooctane-d16 (7 CI, 9 CI), Cyclooctane-D16 |
| InChI Formula | InChI=1 S/C8H16/c1-2-4-6-8-7-5-3-1/h1-8H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2 |
| SMILES | [2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-hexadecadeuteriocyclooctane |
| Molecular Weight (g/mol) | 128.3112 |
| Formula Weight | 128.2256 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction