missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctylamine, 97+%, Thermo Scientific™

Product Code. 11374404
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11374404 25 g 25g
11364404 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11374404 Supplier Thermo Scientific Alfa Aesar Supplier No. L00981.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Cyclooctylamine is used as pharmaceutical intermediates.

Solubility
Soluble in water. 10.2 g/L at 20°C.

Notes
Protect from moisture, heat. Avoid excessive and light exposure. Air sensitive. Store at +15°C to +25°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5452-37-9
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD00001748
InChI Key HSOHBWMXECKEKV-UHFFFAOYSA-N
Synonym cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc
PubChem CID 2903
IUPAC Name cyclooctanamine
SMILES C1CCCC(CCC1)N

Specifications

Melting Point -48°C
Density 0.912
Boiling Point 189°C to 190°C
Flash Point 62°C (143°F)
Odor Amine-like
Refractive Index 1.481
Quantity 25 g
UN Number UN2735
Beilstein 2689120
Sensitivity Air Sensitive
Solubility Information Soluble in water. 10.2g/L at 20°C.
Formula Weight 127.23
Percent Purity ≥97%
Chemical Name or Material Cyclooctylamine
Show More Show Less
compliance-icons
Product Identifier
  • Cyclooctylamine
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.