missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarbonyl Chloride, TRC

Product Code. 30363072
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363072 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363072 Supplier Toronto Research Chemicals Supplier No. TRCC992360100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopentanecarbonyl Chloride
CAS 4524-93-0
Molecular Formula C6H9ClO
Quantity 100 mg
InChI Formula InChI=1S/C6H9ClO/c7-6(8)5-3-1-2-4-5/h5H,1-4H2
SMILES ClC(=O)C1CCCC1
IUPAC Name cyclopentanecarbonyl chloride
Molecular Weight (g/mol) 132.59
Formula Weight 132.0342
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.