missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxaldehyde, 98%, stab.

Product Code. 15422806
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422806 5 g 5g
15412806 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422806 Supplier Thermo Scientific Alfa Aesar Supplier No. H56609.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 872-53-7 | C6H10O | 98.145 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water and ethanol

Notes
Light sensitive. Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-53-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00060798
InChI Key VELDYOPRLMJFIK-UHFFFAOYSA-N
Synonym cyclopentanecarboxaldehyde, cyclopentanealdehyde, cyclopentancarbaldehyd, cyclopentylformaldehyde, formylcyclopentane, cyclopentyl aldehyde, 1-formylcyclopentane, cyclopentane-1-carboxaldehyde, cyclopentanecaboxaldehyde, cyclopentanecarboaldehyde
PubChem CID 70106
IUPAC Name cyclopentanecarbaldehyde
SMILES C1CCC(C1)C=O

Specifications

Density 0.919
Boiling Point 140°C to 141°C
Flash Point 28°C (82°F)
Refractive Index 1.443
Quantity 5 g
UN Number UN1989
Solubility Information Soluble in water and ethanol
Formula Weight 98.14
Percent Purity 98%
Chemical Name or Material Cyclopentanecarboxaldehyde, stabilized with 25ppm 1,4-benzoquinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.