missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanecarboxaldehyde, TRC

Product Code. 30354477
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30354477 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354477 Supplier Toronto Research Chemicals Supplier No. TRCC9894555G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopentanecarboxaldehyde
CAS 872-53-7
Molecular Formula C6H10O
Quantity 5 g
Synonym Cyclopentanealdehyde, Cyclopentanecarbaldehyde, Cyclopentyl aldehyde, Cyclopentylformaldehyde, Formylcyclopentane, NSC 17492
InChI Formula InChI=1S/C6H10O/c7-5-6-3-1-2-4-6/h5-6H,1-4H2
SMILES O=CC1CCCC1
IUPAC Name cyclopentanecarbaldehyde
Molecular Weight (g/mol) 98.14
Formula Weight 98.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.