missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl Phenyl Ketone, TRC

Product Code. 30353005
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30353005 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353005 Supplier Toronto Research Chemicals Supplier No. TRCC9902651G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Cyclopentylphenylmethanone
CAS 5422-88-8
Molecular Formula C12 H14 O
Quantity 1 g
Synonym Cyclopentylphenylmethanone, Glycopyrronium Bromide Imp. M (EP), Benzoylcyclopentane, Cyclopentyl phenyl ketone, NSC 10749, NSC 401765, Phenyl cyclopentyl ketone, Cyclopentyl-phenyl-ketone
InChI Formula InChI=1S/C12H14O/c13-12(11-8-4-5-9-11)10-6-2-1-3-7-10/h1-3,6-7,11H,4-5,8-9H2
SMILES O=C(C1CCCC1)c2ccccc2
IUPAC Name cyclopentyl(phenyl)methanone
Molecular Weight (g/mol) 174.24
Formula Weight 174.1045
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.