Learn More
Cyclophilin B Antibody (CL3915) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59782AF405
Product Code. 30302809
  • 737.00€ / 0.10mL
Description
Cyclophilin B Monoclonal antibody specifically detects Cyclophilin B in Human,Mouse,Rat samples. It is validated for Western Blot,KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cyclophilin B
Monoclonal
Alexa Fluor 405
Cyclophilin B, cyclophilin-like protein, CYPBEC 5.2.1.8, CYP-S1, MGC2224, OI9MGC14109, peptidyl-prolyl cis-trans isomerase B, peptidylprolyl isomerase B (cyclophilin B), PPIase B, Rotamase B, S-cyclophilin, SCYLP
This antibody was developed using a synthetic peptide derived from P23284, with the exact immunogen sequence remaining proprietary.
0.1 mL
Loading Controls
5479
Store at 4⪚C in the dark.
IgG2b
Western Blot, KnockDown
CL3915
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat