missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropanecarbonyl Chloride, TRC

Product Code. 30351626
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30351626 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30351626 Supplier Toronto Research Chemicals Supplier No. TRCC99006050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopropanecarbonyl Chloride
CAS 4023-34-1
Molecular Formula C4H5ClO
Quantity 50 g
Synonym Cyclopropanecarboxylic Acid Chloride, Cyclopropanoyl Chloride, Cyclopropylcarbonyl Chloride, Cyclopropylcarboxylic Acid Chloride
InChI Formula InChI=1S/C4H5ClO/c5-4(6)3-1-2-3/h3H,1-2H2
SMILES ClC(=O)C1CC1
IUPAC Name cyclopropanecarbonyl chloride
Molecular Weight (g/mol) 104.53
Formula Weight 104.0029
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.