missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyl-1-d1-methyl-d2 Alcohol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Cyclopropylmethanol-1,1,1'-D3 |
| CAS | 1219802-62-6 |
| Molecular Formula | C4 D3 H5 O |
| Quantity | 0.25 g |
| Synonym | Cyclopropane-1-d-methan-d2-ol, Cyclopropane-1-d1-methan-α,α-d2-ol Cyclopropyl-1-d1-methyl-d2 Alcohol, Cyclopropylmethanol-1,1,1'-D3 |
| InChI Formula | InChI=1 S/C4H8O/c5-3-4-1-2-4/h4-5 H,1-3H2/i3D2,4 D |
| SMILES | [2 H]C([2 H])(O)C1([2 H])CC1 |
| IUPAC Name | dideuterio-(1-deuteriocyclopropyl)methanol |
| Molecular Weight (g/mol) | 75.1242 |
| Formula Weight | 75.0763 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction