missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylacetonitrile, 97%

Product Code. 11342727
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342727 5 g 5g
11358515 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342727 Supplier Thermo Scientific Alfa Aesar Supplier No. L11950.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6542-60-5 | C5H7N | 81.118 g/mol

Cyclopropylacetonitrile is used in the preparation of cyclopropyl-2 acetophenone and cyclopropylacetyl chloride. It acts as an organic intermediate in organic synthesis. It is also employed in the preparation of agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylacetonitrile is used in the preparation of cyclopropyl-2 acetophenone and cyclopropylacetyl chloride. It acts as an organic intermediate in organic synthesis. It is also employed in the preparation of agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Miscible with water, methylene chloride and alcohols.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6542-60-5
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00041523
InChI Key FAUQRRGKJKMEIW-UHFFFAOYSA-N
Synonym cyclopropylacetonitrile, cyclopropaneacetonitrile, 2-cyclopropylethanenitrile, cyclopropyl acetonitrile, cyclopropyl-acetonitrile, acmc-20albr, cyanomethyl cyclopropane, ksc352m6t
PubChem CID 643447
IUPAC Name 2-cyclopropylacetonitrile
SMILES C1CC1CC#N

Specifications

Density 0.89
Boiling Point 142°C to 144°C
Flash Point 43°C (109°F)
Odor Strong
Refractive Index 1.4245
Quantity 5 g
UN Number UN3275
Beilstein 2036964
Solubility Information Miscible with water,methylene chloride and alcohols.
Formula Weight 81.12
Percent Purity 97%
Chemical Name or Material Cyclopropylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.