missing translation for 'onlineSavingsMsg'
Learn More

Cyhalofop, TRC

Product Code. 30352492
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352492 50 mg 50mg
30354943 500 mg 500mg
30349613 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30352492 Supplier Toronto Research Chemicals Supplier No. TRCC93725050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122008-78-0
Molecular Formula C16 H12 F N O4
Molecular Weight (g/mol) 301.27
Synonym Propanoic acid, 2-[4-(4-cyano-2-fluorophenoxy)phenoxy]-, (R)-, (2R)-2-[4-(4-Cyano-2-fluorophenoxy)phenoxy]propanoic acid, (R)-(+)-2-[4-(2-Fluoro-4-cyanophenoxy)phenoxy]propanoic acid, Cyhalofop, Cyhalofop acid
IUPAC Name (2R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propanoic acid
SMILES C[C@@H](Oc1ccc(Oc2ccc(cc2F)C#N)cc1)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H12FNO4/c1-10(16(19)20)21-12-3-5-13(6-4-12)22-15-7-2-11(9-18)8-14(15)17/h2-8,10H,1H3,(H,19,20)/t10-/m1/s1
Formula Weight 301.075
Chemical Name or Material Cyhalofop
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.