missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CYM 50308
GREENER_CHOICE

Product Code. 30001554
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001554

Brand: Tocris Bioscience™ 4678/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective S1P4 agonist

Potent and selective sphingosine-1-phosphate receptor 4 (S1P4) agonist (EC50 values are 56 nM and 2100 nM for S1P4 and S1P5 receptors, respectively). Displays no activity at S1P1, S1P2 and S1P3 receptors at concentrations up to 25 μM.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1345858-76-5
Molecular Formula C20H21F2N3O2S

Specifications

Quantity 10 mg
Formula Weight 405.46
Purity >98%
Molecular Weight (g/mol) 405.464
Chemical Name or Material CYM 50308
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.