Learn More
R&D Systems™ Cyno Monkey BAFF/TNFSF13B (XP_005586281) VersaClone cDNA
Specifications
Specifications
| Accession Number | XP_005586281 |
| Gene ID (Entrez) | 10673 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |