missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cyno Monkey CRACC/SLAMF7 (XP_005541294) VersaClone cDNA

Product Code. 18726089
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18726089 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18726089 Supplier R&D Systems™ Supplier No. RDC2954
  • ONLINE EXCLUSIVE PRICE
    341.00€
    322.35€ / 10µg
    Save 18.65€
    5% Off
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene CRACC/SLAMF7
Accession Number XP_005541294
Gene ID (Entrez) 57823
Gene Alias 19 A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19 A, SLAM family member 7, SLAMF7
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.