missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Cyno Monkey IFN-alpha/beta R1 (EHH61907) VersaClone cDNA
Description
- IFNAR1 interferon alpha and beta receptor subunit 1 [ Macaca fascicularis (crabeating macaque) ]
- IFN-alpha/beta R1, also known as IFNAR1, is a 120 kDa component of the class II cytokine receptor that contributes to the functional receptor for type I IFNs
- IFN-alpha/beta R1 associates with the 100 kDa IFN-alpha/beta R2 component to form a receptor complex that transmits anti-viral and anti-proliferative signals
- Down-regulation of the receptor is accomplished by ubiquitination and degradation
- Binding and activation of IFNAR1 stimulates Janus protein kinases, which in turn phosphorylate several proteins, including STAT1 and STAT2. It also functions as an antiviral factor
- The IFN-alpha/beta R1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | cynoIFNAR1 |
| Accession Number | EHH61907 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1693 bp |
| Gene ID (Entrez) | 3454 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?