missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cynomolgus Monkey BCMA (XP_005591343) VersaClone cDNA

Product Code. 18735239
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18735239 10 μg 10µg
1 options
This item is not returnable. View return policy

Product Code. 18735239

Brand: R&D Systems™ RDC1903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Cynomolgus Monkey BCMA (XP_005591343) VersaClone cDNA is a member of the TNF receptor superfamily.

The Cynomolgus Monkey BCMA (XP_005591343) VersaClone cDNA clone contains the complete ORF for BCMA/TNFRSF17, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The BCMA/TNFRSF17 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifications

Gene cynoTNFRSF17
Accession Number XP_005591343
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 565bp
pH 8.5
Gene ID (Entrez) 608
Gene Alias B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, TNFRSF13A, TNFRSF17, tumor necrosis factor receptor superfamily, member 17
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Cynomolgus Monkey
Quantity 10 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.