Learn More
CYP2W1 Mouse anti-Human, Unconjugated, Clone: 2B10H2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67419-1-IG-20UL
Description
The CYP2W1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CYP2W1. This antibody recognizes human, mouse, rat antigen. The CYP2W1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| CYP2W1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 2B10H2 | |
| Unconjugated | |
| 20 μL | |
| 54905 | |
| IgG1 |