missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP51A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13431-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum protein participates in the synthesis of cholesterol by catalyzing the removal of the 14alpha-methyl group from lanosterol. Homologous genes are found in all three eukaryotic phyla, fungi, plants, and animals, suggesting that this is one of the oldest cytochrome P450 genes. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| CYP51A1 | |
| Polyclonal | |
| Unconjugated | |
| CYP51A1 | |
| CP51, CYP51, CYP51A1, CYPL1, CYPLI, Cytochrome P450 14DM, Cytochrome P450 51A1, Cytochrome P45014DM, Cytochrome P450LI, LDM, P450 14DM, P450L1, Sterol 14 alpha demethylase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13121, 1595, 25427 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunofluorescence | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16850, Q64654, Q8K0C4 | |
| Cyp51a1 | |
| CYP51A1 Fusion Protein Ag4235 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction