Learn More
Riley™ Cypher Eco Safety Glasses
Description
- Sports style 8 Base lens
- Soft, adjustable anti-slip nose bridge
- Co-injected bi-colour temples with flexible TPE ends enhance overall fit and comfort
- No single-use packaging, supplied in a microfiber pouch made from 100% recycled material
- Unique aero-slotted side arms
- Available for prescription
Specifications
Specifications
| Certifications/Compliance | CE or - UKCA or - EN 166:2001 |
| Lens Coating | Anti-fog, Scratch-resistant, Moisture Repellent, UV-treated |
| Lens Tint | Clear |
| Product Type | Safety Glasses |
| For Use With (Application) | Component and Product Assembly, Vehicle Repairs and Maintenance, Commercial Cleaning, General Maintenance, Inspection and Quality Control |
| Frame Color | Black |
| Transmittance Value | 92% |
| Weight (Metric) | 22 g |
| Material | Polycarbonate |
| Disposable | Reusable |
| Show More |